Learn More
DiscoverX Recombinant Human EG-VEGF
Code produit 17332369
Encompass
Change view
Click to view available options
:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit
17332369 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17332369 Fournisseur DiscoverX Code fournisseur 921022
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement
Recombinant Human EG-VEGF is an agonist for PKR1 and PKR2. Provided as 5μg of dry powder alongside reconstitution buffer.
EG-VEGF (PK1) is a prokinectin growth factor expressed in the ovaries, testis, adrenal, and placental tissues, and is one of the main non-glycosylated proteins in the body. It acts as an angiogenic mitogen and has chemotactic function. EG-VEGF acts as an agonist on PKR1 and PKR2. Provided as 5μg of dry powder alongside reconstitution buffer.
Spécification
Quantité 5 μg
Type de produit Control Ligands