missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAB2B Protein

Code produit 15982929
Change view
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15982929 20 μg 20µg
15992929 5 μg 5µg
15972929 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15982929 Fournisseur enQuireBio™ Code fournisseur QP1323620ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

A cDNA sequence encoding the RAB2B was constructed and used to recombinantly synthesize the protein.

Specifications

Nom RAB2B Protein
Quantité 20 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MGSHMTYAYL FKYIIIGDTG VGKSCLLLQF TDKRFQPVHD LTIGVEFGAR MVNIDGKQIK LQIWDTAGQE SFRSITRSYY RGAAGALLVY DITRRETFNH LTSWLEDARQ HSSSNMVIML IGNKSDLESR RDVKREEGEA FAREHGLIFM ETSAKTACNV EEAFINTAKE IYRKIQQGLF DVHNEANGIK IGPQQSISTS VGPSASQRNS RDIGSNSGCC
Tampon RAB2B protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.15M NaCl.
Pureté ou qualité Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.