missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RND1 Protein

Code produit 15983219
Change view
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15983219 20 μg 20µg
15993219 5 μg 5µg
15973219 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15983219 Fournisseur enQuireBio™ Code fournisseur QP1332720ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

A cDNA sequence encoding the RND1 was constructed and used to recombinantly synthesize the protein.

Specifications

Nom RND1 Protein
Quantité 20 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MKERRAPQPV VARCKLVLVG DVQCGKTAML QVLAKDCYPE TYVPTVFENY TACLETEEQR VELSLWDTSG SPYYDNVRPL CYSDSDAVLL CFDISRPETV DSALKKWRTE ILDYCPSTRV LLIGCKTDLR TDLSTLMELS HQKQAPISYE QGCAIAKQLG AEIYLEGSAF TSEKSIHSIF RTASMLCLNK PSPLPQKSPV
Tampon The RND1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 30% glycerol.
Pureté ou qualité Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.