Learn More
Contec Replacement Collars
Code produit 31777564 Tous les produits Contec Produits
Modifier l'affichage
Click to view available options
Comprend:
1 Collar Pole
3 Collar Pole
5 Collar Pole
Conditionnement:
1 pièce
3 pièces
5 pièces
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Comprend Conditionnement
31777564 5 Collar Pole 5 pièces
31777561 1 Collar Pole 1 pièce
31777563 3 Collar Pole 3 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 31777564 Fournisseur Contec Code fournisseur L334
  • ONLINE EXCLUSIVE PRICE
    43.22€
    31.89€ / 5 pièces
    Économisez 11.33€
    26% de réduction
Expédition estimée: 24-03-2027
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass. Compatible with all QuickConnect™ frames Multiple lengths available Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles. Telescoping handles are available in lightweight carbon fiber, as well as anodised aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
  • Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments.
  • Allows investment in one handle with various interchangeable heads required for specific applications.
  • Handles are available in several lengths and styles.
  • Telescoping handles are available in lightweight carbon fiber, anodised aluminum, durable stainless steel, or fiberglass.
  • Choose from a variety of lengths to suit different cleaning needs.
TRUSTED_SUSTAINABILITY
Spécification
Type de produit Cleanroom Mop Handle
Stérilité Non-sterile
Autoclavable Autoclavable
Comprend 5 Collar Pole
Color-coded No
Matériau Carbon Fiber