Learn More
BD Rhapsody™ Intracellular AbSeq Buffer Kit
Code produit 32226987 Tous les produits BD Biosciences Produits
Modifier l'affichage
Click to view available options
Type de produit:
Intracellular AbSeq Buffer Kit
Conditionnement:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de produit Conditionnement
32226987 Intracellular AbSeq Buffer Kit 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 32226987 Fournisseur BD Code fournisseur 570742
 Article restreint
Expédition estimée: 18-11-2026
The BD Rhapsody™ Intracellular AbSeq Buffer Kit allows users to stain for intracellular antigens using intracellular BD™ AbSeq Antibody-Oligos for profiling with the BD Rhapsody™ System.
Each intracellular BD™ AbSeq Antibody is conjugated to an antibody-specific oligonucleotide barcode for profiling alongside surface protein and mRNA expression. This product provides reagents to preserve, permeabilize, and stain cells with intracellular BD™ AbSeq Antibody-Oligos.
Prior to beginning the intracellular CITE-seq protocol, surface antigen staining should be performed using one of the following protocols:
  • BD Rhapsody™ System Single-Cell Labeling with BD™ AbSeq Ab-Oligos (1 plex to 40 plex) Protocol 23-24262 or
  • BD Rhapsody™ System Single-Cell Labeling with the BD™ Single-Cell Multiplexing Kit and BD™ AbSeq Ab-Oligos 23-21339 to support single-cell labeling with BD™ AbSeq Ab-Oligos without and with sample multiplexing, respectively
These protocols have been tested for use with up to 40 surface markers. The following protocols, BD Rhapsody™ System Single-Cell Labeling with BD™ AbSeq Ab-Oligos (41 plex to 100 plex) protocol, 23-22314 or BD Rhapsody™ System Single-Cell Labeling with BD™ Single-Cell Multiplexing Kit and BD™ AbSeq Ab-Oligos protocol, 23-22354, are available for use with high-plex experiments, however, additional optimization may be required.
TRUSTED_SUSTAINABILITY
Spezifikation
Type de produit Intracellular AbSeq Buffer Kit
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.