Learn More
STEMCELL Technologies RoboSep™ Mouse CD138 Positive Selection Kit
Description
- The EasySep™ Mouse CD138 Positive Selection Kit is designed to isolate CD138+ cells from mouse splenocytes, lymph nodes and bone marrow by immunomagnetic positive selection
- Desired cells are targeted with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
- Unwanted cells are simply poured off, while desired cells remain in the tube
- Isolated cells are immediately available for downstream applications such as flow cytometry, cell culture, and hybridoma generation
Spécification
Spécification
| Type de cellule | B Cells, Plasma |
| À utiliser avec (application) | Cell Isolation |
| Quantité | 2 x 109 cellules |