Learn More
STEMCELL Technologies RoboSep™ Tip Head Polishing Compound
Code produit 17118021
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantité:
7 mL
Conditionnement:
7mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
17118021 7 mL 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17118021 Fournisseur STEMCELL Technologies Code fournisseur 20119
 Article restreint
Expédition estimée: 20-11-2026
RoboSep™-S is the next-generation RoboSep™ instrument for fully automated cell separation
  • RoboSep™-S is the next-generation RoboSep™ instrument for fully automated cell separation
  • Using EasySep™ technology, RoboSep™-S performs all steps necessary to magnetically label and separate virtually any cell type by positive or negative selection
  • RoboSep™-S is designed to minimize sample handling, eliminate cross-contamination, and reduce hands-on time
  • Leasing options and warranty coverage are available
Spécification
Quantité 7 mL