Learn More
STEMCELL Technologies RosetteSep™ DM-M Density Medium
Code produit 17137711
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantité:
100 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
17137711 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17137711 Fournisseur STEMCELL Technologies Code fournisseur 15725
 Article restreint
Expédition estimée: 25-11-2026
Density gradient medium for use with RosetteSep™ myeloid cell enrichment cocktail - 100 mL
  • RosetteSep™ DM-M is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human myeloid (CD33+) cells from whole blood.
  • RosetteSep™ DM-M has a density of 1.085 g/mL.
Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled For Research Use Only. Please note there have been no changes to the form, function and quality assurances associated with these products.
Spécification
Quantité 100 ml
Type de cellule Myeloid Cells
À utiliser avec (application) Cell Isolation; In Vitro Diagnostic