Learn More
Invitrogen™ Human RPS6KA2 (RSK3), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. The purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ Assay technologies to identify novel kinase inhibitors. RPS6KA2 is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen & Z'-LYTE assay technology Available in multiple pack sizes for convenient use Also available in our SelectSceen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for RPS6KA2 (RSK3) is NP_066958.2.
Spécification
Spécification
| Numéro d’adhésion | Q15349 |
| Concentration | See Label |
| À utiliser avec (application) | Kinase Assay |
| Formule | 50mM tris HCl with 150mM NaCl, 0.1mM EDTA, 0.25mM DTT, 10mM glutathione, 25% glycerol, 0.1mM PMSF and no preservative; pH 7.5 |
| Identification génétique (Entrez) | 6196 |
| Poids moléculaire | 109.9 kDa |
| Nom | Human RPS6KA2 (RSK3), GST Tag |
| Plage de pH | 7.5 |
| Méthode de purification | Purified |
| Quantité | 10 μg |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.