Learn More
(S)-5-Hydroxy Propafenone Hydrochloride, TRC
Code produit 30403080
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30403080 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30403080 Fournisseur Toronto Research Chemicals Code fournisseur TRCH9524601MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau (S)-5-Hydroxy Propafenone Hydrochloride
CAS 158080-71-8
Formule moléculaire C21 H27 N O4 . Cl H
Quantité 1 mg
Formule InChI InChI=1S/C21H27NO4.ClH/c1-2-12-22-14-18(24)15-26-21-11-9-17(23)13-19(21)20(25)10-8-16-6-4-3-5-7-16;/h3-7,9,11,13,18,22-24H,2,8,10,12,14-15H2,1H3;1H/t18-;/m0./s1
SMILES Cl.CCCNC[C@H](O)COc1ccc(O)cc1C(=O)CCc2ccccc2
Nom IUPAC 1-[5-hydroxy-2-[(2S)-2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one;hydrochloride
Poids moléculaire (g/mol) 393.9
Poids de la formule 393.1707