Learn More
MedChemExpress (S,R,S)-AHPC-PEG2-N3
Code produit 16490086
Encompass_Preferred
Change view
Click to view available options
Quantité:
100 mg
500 mg
1 g
2 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16490086 2 g
16471556 500 mg
16431009 1 g
16424906 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16490086 Fournisseur MedChemExpress Code fournisseur HY103599
MedChemExpress (S,R,S)-AHPC-PEG2-N3 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 2-unit PEG linker used in PROTAC technology.
Spécification
Poids moléculaire 601.72 g/mol
CAS 2010159-45-0
Contenu et stockage -20°C, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (stored under nitrogen)
Conditions d’expédition Room temperature
Quantité 2 g
Synonyme VH032-PEG2-N3 ∣ VHL Ligand-Linker Conjugates 6 ∣ E3 ligase Ligand-Linker Conjugates 13
Solubilité Ethanol : 100 mg/mL (166.19 mM; Need ultrasonic) ∣ DMSO : 50 mg/mL (83.10 mM; Need ultrasonic)