Learn More
Sacubitril/Valsartan, MedChemExpress
Code produit. 16487478
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
50 mg
Code produit. 16487478
Marque: Medchem Express HY18204A
MedChemExpress Sacubitril/Valsartan (LCZ696), comprised Valsartan and Sacubitril (AHU377) in 1:1 molar ratio, is a first-in-class, orally bioavailable, and dual-acting angiotensin receptor-neprilysin (ARN) inhibitor for hypertension and heart failure. Sacubitril/Valsartan ameliorates diabetic cardiomyopathy by inhibiting inflammation, oxidative stress and apoptosis.
Sacubitril/Valsartan (LCZ696), comprised Valsartan and Sacubitril (AHU377) in 1:1 molar ratio, is a first-in-class, orally bioavailable, and dual-acting angiotensin receptor-neprilysin (ARN) inhibitor for hypertension and heart failure. Sacubitril/Valsartan ameliorates diabetic cardiomyopathy by inhibiting inflammation, oxidative stress and apoptosis.
Spécification
Nom chimique ou matériau Sacubitril/Valsartan
Forme physique Powder
CAS 936623-90-4
Couleur Blanc
Quantité 10 mg
Notes de qualité de la pureté Research
Formule moléculaire C48H55N6Na3O8.2.5H2O
À utiliser avec (application) COVID-19-immunoregulation
Synonyme LCZ696
Informations sur la solubilité DMSO : ≥ 100 mg/mL (104.39 mM) ∣H2O : ≥ 50 mg/mL (52.19 mM)
SMILES O=C(CCC(O[Na])=O)N([C@H](CC(C=C1)=CC=C1C2=CC=CC=C2)C[C@@H](C)C(OCC)=O)[H].O=C(O[Na])[C@@H](N(C(CCCC)=O)CC3=CC=C(C4=CC=CC=C4C5=NN=NN5[Na])C=C3)C(C)C.[2.5H2O]
Poids moléculaire (g/mol) 957.99
Poids de la formule 957.99
Pourcentage de pureté 99.0%
Qualité Research
Afficher plus Afficher moins
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.