Learn More
SAH3 Antibody, Allophycocyanin, R&D Systems™
Product Code. 18609791 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18609791 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18609791 Supplier Novus Biologicals Supplier No. NBP298616APC
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
SAH3 Polyclonal specifically detects SAH3 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Gene Alias adenosylhomocysteinase-like 2, AdoHcyase 3, ADOHCYASE3, EC 3.3.1.1, FLJ21719, KIAA0828S-adenosylhomocysteine hydrolase-like 2, putative adenosylhomocysteinase 3, S-adenosylhomocysteine hydrolase-like protein 2, S-adenosyl-L-homocysteine hydrolase 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SAH3 fragment.
Gene ID (Entrez) 23382
Product Type Antibody
Isotype IgG
Product Line Novus