Learn More
Thermo Scientific™ Sample Storage Starter Packs
Code produit 17896148
Modifier l'affichage
Click to view available options
Comprend:
Tubes de 0,5 ml, déboucheuse, lecteur
Tubes de 1,0 ml, déboucheuse, lecteur
Tubes et déboucheuse
Tubes, déboucheuse, lecteur HSX
Tubes, déboucheuse, lecteur ST
Conditionnement:
1 kit
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Comprend Conditionnement
17896148 Tubes de 0,5 ml, déboucheuse, lecteur 1 kit
17886148 Tubes et déboucheuse 1 kit
17866148 Tubes, déboucheuse, lecteur ST 1 kit
17876148 Tubes, déboucheuse, lecteur HSX 1 kit
17856148 Tubes de 1,0 ml, déboucheuse, lecteur 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 17896148 Fournisseur Thermo Scientific™ Code fournisseur HHDHSXWP1D505
  • ONLINE EXCLUSIVE PRICE
    17260.00€
    15098.69€ / 1 kits
    Économisez 2161.31€
    12% de réduction
 Article restreint
Expédition estimée: 27-11-2026
Règlementaire : Cet article est soumis à éco-contribution.
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
  • Choose from five different starter pack options.
  • Eliminate the need to reformat non-2D cryotube samples.
  • Secure sample tracking with 2D, 1D, and human readable codes.
  • Cap/decap a full rack of tubes in less than 1 minute.
  • Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
  • Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
TRUSTED_SUSTAINABILITY
Spécification
Comprend Tubes de 0,5 ml, déboucheuse, lecteur
Type Premium 0.5 mL Starter Pack