Learn More
SB-408124 Hydrochloride, MedChemExpress
Code produit 16480648
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16480648 5 mg
16408218 10 mg
16407956 100 mg
16404117 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16480648 Fournisseur Medchem Express Code fournisseur HY76612
MedChemExpress SB-408124 Hydrochloride is a selective non-peptide orexin receptor 1 (OX1) receptor antagonist with Kis of 57 nM and 27 nM in whole cell and membrane, respectively. SB-408124 Hydrochloride exhibits 50-fold selectivity over OX2 receptor.
SB-408124 Hydrochloride is a selective non-peptide orexin receptor 1 (OX1) receptor antagonist with Kis of 57 nM and 27 nM in whole cell and membrane, respectively. SB-408124 Hydrochloride exhibits 50-fold selectivity over OX2 receptor.
Spécification
Nom chimique ou matériau SB-408124 Hydrochloride
CAS 1431697-90-3
Couleur Gris
Notes de qualité de la pureté Research
Formule moléculaire C19H19ClF2N4O
Quantité 5 mg
Informations sur la solubilité H2O : 1 mg/mL (2.55 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C(NC1=CC=C(C=C1)N(C)C)NC2=CC(C)=NC(C2=CC(F)=C3)=C3F.[H]Cl
Poids moléculaire (g/mol) 392.83
Poids de la formule 392.83
Pourcentage de pureté 98.09%
Qualité Research
Forme physique Solid
À utiliser avec (application) Neuroscience-Neuromodulation