Learn More
Beschreibung
Perform accurate AST with Staphylococcus species specific formulary as part of EU Surveillance program with Thermo Scientific™ Sensititre™ EU Surveillance Staphylococcus EUST2 AST Plate.
This updated format for monitoring Methicillin-resistant Staphylococcus aureus isolates is designed based on EU surveillance program specifications1. Sensititre plates utilise high-quality, ready to use AST broth microdilution (BMD) method which is the gold standard2 phenotypic method for Antimicrobial Susceptibility Testing (AST).
Comply with EU Surveillance program on antimicrobial resistance for Staphylococcus species, specifically Methicillin-resistant S.aureus strains, with Sensititre EU Surveillance Staphylococcus EUST2 AST Plate 1
- Extended shelf life, room temperature storage eliminates inventory control concerns
- Individual packaging allows a laboratory to test one plate at a time with no waste
- Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
Antimicrobial Resistance (AMR) is a significant concern and of increasing importance. The inappropriate use of antimicrobial agents in humans and animals is leading this growing and dangerous issue, emerging as a public health threat. This is why the European Parliament, the Council and its Agencies: European Centre for Disease Prevention and Control (ECDC), the European Food Safety Authority (EFSA) and the European Medicines Agency (EMA), have identified the need to establish a common strategy in the fight against AMR.
The Sensititre System utilizes true minimum inhibitory concentration (MIC) results, which are essential to identifying antimicrobial resistance, and provide greater sensitivity for better resistance tracking.
With a wide range of Sensititre plates specifically designed for surveillance programs, the Sensititre System is a leader in antimicrobial susceptibility testing for surveillance.
Not all products are available for sale in all territories. Please inquire.
Spezifikation
Spezifikation
| Description | Sensititre™ EU Surveillance Staphylococcus EUST2 AST Plate |
| Quantité | 10/Box |
| Options de lecture | Manual, Vizion |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.