Learn More
Thermo Scientific™ Sensititre™ Manual Viewbox
Code produit 16209121
Modifier l'affichage
Click to view available options
Type de produit:
Manual Viewbox
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de produit Conditionnement
16209121 Manual Viewbox 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16209121 Fournisseur Thermo Scientific™ Code fournisseur YV4007
  • ONLINE EXCLUSIVE PRICE
    703.78€
    618.14€ / 1 pièces
    Économisez 85.64€
    12% de réduction
Expédition estimée: 22-12-2026
Perform simple visual reads of your 96-well microtitre plates with the mirrored Thermo Scientific™ Sensititre™ Manual Viewbox.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Spécification
Description Sensititre Manual Viewbox
Type de produit Manual Viewbox
Type de test Minimum Inhibitory Concentration (MIC)