missing translation for 'onlineSavingsMsg'
Learn More

SHE Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22902-1-AP-150UL

Nacres classification code: NA.46

 View more versions of this product

Product Code. 16892125

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody specifically recognizes the 54 kDa protein.

SHE, also known as SH2 domain-containing adapter protein E, is a 495 amino acid protein whose molecular weight is 54 kDa. The function of SHE remains unknown.
Specifications

Specifications

SHE
Polyclonal
Unconjugated
SHE
RP11 350G8.8, SHE
Rabbit
Antigen Affinity Chromatography
RUO
126669, 214547
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5VZ18, Q8BSD5
She
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.