Learn More
Thermo Scientific™ Single-Use BioProcess Container, 2D, Aegis™ 5-14, 2 ports
GREENER_CHOICE
Code produit 17859776 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Capacité (métrique):
50 ml
100 ml
250 ml
500 ml
1 l
2 l
3 l
Conditionnement:
10 pièces
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Capacité (métrique) unitSize
17859776 3 l 10 pièces
17899756 50 ml 10 pièces
17809696 100 ml 10 pièces
17829766 250 ml 10 pièces
17839766 500 ml 10 pièces
17899686 1 l 10 pièces
17849776 2 l 10 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Code produit 17859776 Fournisseur Thermo Scientific™ Code fournisseur SH31265.17
Please to purchase this item. Need a web account? Register with us today!
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
Features:
• All BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Flexible and economic solution to update or increase your bioreactor capacity
• Suitable for both suspension and adherent cultures
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from a large component library – with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and gas filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.
TRUSTED_SUSTAINABILITY
Spécification
Capacité (métrique) 3 l
Quantité 3000 mL
Tubing Style TPE