Learn More
Thermo Scientific™ Single-Use BioProcess Container, 2D, Aegis™ 5-14, 2 ports
Product Code. 17899756 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacité (métrique):
50 ml
100 ml
250 ml
500 ml
1 l
2 l
3 l
Unit Size:
10 pièces
30 pièces
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacité (métrique) Unit Size
17899756 50 ml 10 pièces
17809696 100 ml 10 pièces
17829766 250 ml 10 pièces
17839766 500 ml 10 pièces
17899686 1 l 10 pièces
17849776 2 l 10 pièces
30040086 2 l 30 pièces
17859776 3 l 10 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17899756 Supplier Thermo Scientific™ Supplier No. SH31265.11
  • ONLINE EXCLUSIVE PRICE
    170.00€
    132.35€ / 10 pièces
    Économisez 37.65€
    22% de réduction
 Article restreint
Expédition estimée: 26-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
Features:
  • All BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
  • Manufactured with industry-leading Aegis 5-14 film
  • C-flex tubing that can be used with tubing welder and various connection options
  • Flexible and economic solution to update or increase your bioreactor capacity
  • Suitable for both suspension and adherent cultures
  • Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from a large component library – with over 2,000 unique components that include:
  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
  • Filters—used for bioburden reduction; sterilization grade filters and gas filters.
  • Tubing—thermoplastic elastomers, platinum-cured silicone.
  • Sample ports—septum-style and needle-free ports.
  • Clamps—sliders, pinch, and crimp bands.
TRUSTED_SUSTAINABILITY
Specifications
Concept écologique ISCC PLUS
Capacité (métrique) 50 ml
Quantité 50 mL
Tubing Style TPE