Learn More
Sodium Hydroxide Pearls, Analytical Grade, Penta Chemicals
Description
About us
The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.
We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.
High quality and flexibility
Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.
We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.
Environmental responsibility
Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.
Spécification
Spécification
| Nom chimique ou matériau | Sodium Hydroxide |
| CAS | 1310-73-2 |
| Quantité | 1000 g |
| Pourcentage de pureté | ≥98% |
| Qualité | Analytique |
| Conditionnement | Plastic Jar |
| Forme physique | Pearls |
Sécurité et traitement
- Sodium hydroxide pearls
- Dangers
- Lésion oculaire grave/irritation oculaire Catégorie 1
- Substances et mélanges corrosifs pour les métaux Catégorie 1
- Corrosion cutanée/irritation cutanée Catégorie 1A
- H290-Peut être corrosif pour les métaux.
- H314-Provoque des brûlures de la peau et des lésions oculaires graves.
- H318-Provoque des lésions oculaires graves.
- P280-Porter des gants de protection/des vêtements de protection/un équipement de protection des yeux/ du visage.
- P305+P351+P338-EN CAS DE CONTACT AVEC LES YEUX: rincer avec précaution à l'eau pendant plusieurs minutes. Enlever les lentilles de contact si la victime en porte et si elles peuvent être facilement enlevées. Continuer à rincer.
- P310-Appeler immédiatement un CENTRE ANTIPOISON/un médecin/ .
- MIXTURE LIST-Contain: Sodium hydroxide
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.