Learn More
Sodium Starch Glycolate, TRC
Code produit 30133810
Change view
Click to view available options
Quantité:
100 mg
1 g
5 g
Conditionnement:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30133810 1 g 1g
30132743 100 mg 100mg
17997714 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30133810 Fournisseur Toronto Research Chemicals Code fournisseur TRCS6669901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 9063-38-1
Synonyme Carboxymethyl Starch Sodium Salt, Covagel, Crayon B 300P, Deprogel, Emsize CMS 100, Emsize CMS 60, Estarl A 100, Explosol, Explotab, Explotab CL-V, Expotab PCF, F 500 Papeal No. 50, Glycol YS, Glycolys LV, Glycolys Std, KMK-BUR 1, KMK-OK, Kiprogum F 500, LCW Covagel, Libeng, P 5000, Papeal F 500 No. 50, Polvitex Z, Polytex 60, Primojel, Sodium CM-starch, Sodium Carboxymethyl Starch, Solvitose CL, Vivastar P, Vivastar P 3500, Vivastar P 5000, WS 1R, Walocel CRT 100GA, USP Sodium Starch Glycolate Type A
Spécification
Nom chimique ou matériau Sodium Starch Glycolate Type A
Quantité 1 g