Learn More
Sofosbuvir impurity D, MedChemExpress
Code produit 16471248
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantité:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16471248 5 mg
16432469 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16471248 Fournisseur Medchem Express Code fournisseur HYI0723
 Article restreint
Expédition estimée: 22-09-2026
MedChemExpress Sofosbuvir impurity D is an impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.
Sofosbuvir impurity D is an impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.
Spécification
Nom chimique ou matériau Sofosbuvir impurity D
Couleur Light Yellow
Notes de qualité de la pureté Research
Formule moléculaire C22H29FN3O9P
Quantité 5 mg
SMILES O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@H]1[C@H](O)[C@@](F)(C)[C@@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
Poids moléculaire (g/mol) 529.45
Poids de la formule 529.45
Qualité Research
Forme physique Solid