Learn More
Molecular Devices SpectraMax™ Mini AF System with SoftMax Pro Software
Code produit 31936333
Modifier l'affichage
Click to view available options
Software Type:
SoftMax Pro
Conditionnement:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Software Type Conditionnement
31936333 SoftMax Pro 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 31936333 Fournisseur Molecular Devices Code fournisseur SMAX MINI AF
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
Règlementaire : Cet article est soumis à éco-contribution.
Perform fluorescence‐based detection for microplate‐based assays in a compact format. Supports accessible, multi‐mode screening workflows for routine applications.
  • Designed for fluorescence-based detection workflows in microplate assays
  • Supports routine screening applications across life science workflows
  • Provides a compact microplate reader solution for flexible laboratory environments
  • Enables easy assay setup with configurable detection parameters
  • Supports microplate-based workflows for biochemical and cell-based assays
  • Includes preconfigured protocols to support common assay applications
  • Integrated with SoftMax Pro Software for data acquisition and analysis
  • Designed for ease of use and accessible operation in research laboratories
TRUSTED_SUSTAINABILITY
Spécification
Température de l’incubateur 4-Zone Incubation to 45°C
Vitesse de mesure 11 sec. (96 wells), 22 sec. (384 wells)
Agitation Linear, Orbital
Type de plaques 6 to 384-well Plates
Type de détecteur PMT
Software Type SoftMax Pro
Source lumineuse Xenon Flash
Reading Options Endpoint, Kinetic