Learn More
Labconco™ SpillStopper™ Work Surface for 6 Ft. Protector™Premier™and Protector™XL Hoods
Code produit 17455902 Tous les produits Labconco Corporation Produits
Modifier l'affichage
Click to view available options
Comprend:
Right Rear
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Comprend Conditionnement
17455902 Right Rear 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17455902 Fournisseur Labconco™ Code fournisseur 9500601
  • ONLINE EXCLUSIVE PRICE
    2090.00€
    1744.08€ / 1 pièces
    Économisez 345.92€
    16% de réduction
 Article restreint
Expédition estimée: 25-01-2027
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow in the hood
  • Pre-drilled holes for venting and plumbing passthroughs provided
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 6 x 3 in. (15.2 x 7.6cm) cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
9500622
Not for use with Protector Solvent Storage Cabinets
Spécification
À utiliser avec (équipement) Protector XL Laboratory Hood
Comprend Right Rear
Type de produit Work Surface
Matériau Epoxy Resin
À utiliser avec (application) Laboratory Fume Hood Work