Learn More
SRPR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12090-1-AP-20UL
Nacres classification code: NA.46
 View more versions of this product
Product Code. 16899593
  • 145.00€ / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
The gene encodes a subunit of the endoplasmic reticulum signal recognition particle receptor that, in conjunction with the signal recognition particle, is involved in the targeting and translocation of signal sequence tagged secretory and membrane proteins across the endoplasmic reticulum. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
SRPR
Polyclonal
Unconjugated
SRPRA
Docking protein alpha, DP, DP alpha, SR alpha, Sralpha, SRP alpha, SRPR, SRPR alpha
Rabbit
Antigen Affinity Chromatography
RUO
315548, 6734, 67398
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08240, Q9DBG7
Srpr, Srpra
SRPR Fusion Protein Ag2730
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates