Learn More
SSE15206, MedChemExpress
Code produit 16493086
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16493086 25 mg
16484448 10 mg
16435336 100 mg
16427177 5 mg
16420767 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Code produit 16493086 Fournisseur Medchem Express Code fournisseur HY111425
MedChemExpress SSE15206 is a microtubule polymerization inhibitor (GI50 = 197 nM in HCT116 cells) that overcomes multidrug resistance. Causes aberrant mitosis resulting in G2/M arrest due to incomplete spindle formation in cancer cells.
SSE15206 is a microtubule polymerization inhibitor (GI50 = 197 nM in HCT116 cells) that overcomes multidrug resistance. Causes aberrant mitosis resulting in G2/M arrest due to incomplete spindle formation in cancer cells.
Spécification
Nom chimique ou matériau SSE15206
CAS 1370046-40-4
Couleur Light Yellow
Notes de qualité de la pureté Research
Formule moléculaire C19H21N3O3S
Quantité 25 mg
Informations sur la solubilité DMSO : 150 mg/mL (403.82 mM; Need ultrasonic)
SMILES COC1=C(OC)C(OC)=CC(C2N(C(N)=S)N=C(C3=CC=CC=C3)C2)=C1
Poids moléculaire (g/mol) 371.45
Poids de la formule 371.45
Pourcentage de pureté 98.39%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death