Learn More
Poudre d’acier inoxydable, maille de -100, Type304-L, Thermo Scientific Chemicals
Description
Stainless Steel powder, -100 mesh, Type 304-L is the most common stainless steel alloy widely used in wire weaving. It has excellent corrosion resistance and can withstand temperatures of 1100 degrees F. It has better welding characteristics for lower carbon content.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Nom chimique ou matériau | Stainless Steel powder, Type 304-L |
| Moy. Mol. Poids ou mol. Plage de poids | Fe:Cr:Ni; 70:19:11 wt% |
| Quantité | 50 g |
| Dimensions de la maille | −100 Mesh |
| Forme | Poudre |
| Numéro MDL | MFCD00148499 |
| Informations sur la solubilité | Insoluble in water. |