Learn More
STEMCELL Technologies STEMdiff™ SMADi Neural Induction Kit
Code produit 16620452
Encompass
Change view
Click to view available options
Quantité:
1 kit
Conditionnement:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16620452 1 kit 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16620452 Fournisseur STEMCELL Technologies Code fournisseur 08581
Serum-free medium kit for highly efficient SMAD inhibition-mediated neural induction of human ES and iPS cells
  • STEMdiff™ SMADi Neural Induction Kit consists of a defined, serum-free medium and supplement for the highly efficient neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • This kit combines STEMdiff™ Neural Induction Medium (Catalog #05835) with STEMdiff™ SMADi Neural Induction Supplement, which directs differentiation by blocking TGF-β/BMP-dependent SMAD signaling, resulting in efficient neural induction of even hard-to-differentiate cell lines
  • Neural progenitor cells (NPCs) can be generated using STEMdiff™ SMADi Neural Induction Kit with either an embryoid body (EB) protocol or monolayer culture protocol
  • The resulting cultures are enriched for central nervous system (CNS)-type NPCs, which express SOX1, Nestin, and PAX6
  • NPCs generated using this kit can be passaged as single cells and expanded in STEMdiff™ Neural Progenitor Medium (Catalog #05833)
  • The NPCs can also be differentiated into neurons and glia
Spécification
Type de cellule Neural Cells, PSC-Derived, Pluripotent Stem Cells
À utiliser avec (application) Cell Differentiation
Quantité 1 kit