Learn More
Kern Sohn Microscope à zoom stéréo OZM
Code produit 17359541
Encompass_Preferred
Modifier l'affichage
Click to view available options
Type de tête:
Binoculaire
Trinoculaire
Conditionnement:
1 pièce
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de tête unitSize
17359541 Binoculaire 1 pièce
17369541 Trinoculaire 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 17359541 Fournisseur Kern Sohn Code fournisseur OZM 542
 Article restreint
Expédition estimée: 29-07-2026
Ajouter au panier
Ajouter au panier
Règlementaire : Cet article est soumis à éco-contribution.
First-class optics and strong illumination combined with a high level of flexibility.
  • The KERN OZM-5 series is a range of excellent stereo zoom microscopes with above-average optical features.
  • The ergonomic shape allows a simple, effortless working over a period of several hours.
  • The extraordinarily strong and continuously dimmable 3W LED incident and transmitted illumination ensures a flexible and particularly good level of illumination for your sample.
  • With its large working distance, an extra large field of view and its brilliant resolution, the KERN OZM provides sharp, high-contrast, colour-true images.
  • The zoom objective offers you continuous magnification from 7×–45×.
  • The pillar stand is particularly flexible due to its variable and sturdy adjustment mechanism and therefore enables ergonomic working procedures.
  • A large selection of eyepieces, (universal) stands, a darkfield kit, external illumination units as well as auxiliary objectives and more are available as accessories
  • A protective dust cover, eye cups, as well as multi-lingual user instructions are included in the scope of delivery.
  • Scope of application: in vitro fertilisation, detection of parasites, zoology and botany, tissue preparation, section, quality control, Electronics and semiconductor industry, assembly and repair
  • Applications/Samples: Samples with focus on three-dimensional impression, zoom with variable magnification (depth, thickness), e.g. insects, seeds, circuit boards, components
Spécification
Tension 230 V
Pouvoir grossissant 0.7X to 4.5X
Type de tête Binoculaire
Type d’éclairage LED