Learn More
Phenomenex™ Strata™ C18-E On-Line SPE Column
Code produit 31745408 Tous les produits Phenomenex Produits
Modifier l'affichage
Click to view available options
Granulométrie:
20 μm
50 μm
55 μm
Conditionnement:
1 pièce
4 pièces
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Granulométrie Conditionnement
31745408 20 μm 1 pièce
31742121 55 μm 1 pièce
31743561 50 μm 4 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 31745408 Fournisseur Phenomenex™ Code fournisseur 00MS039B0CB
 Article restreint
Expédition estimée: 28-10-2026
Strata SPE products are manufactured to bring you optimal quality, reproducibility, and performance. With over 20 years of experience, Strata SPE products provide clean extracts and high recoveries for a multitude of target analytes across diverse sample matrices.
  • Available retention mechanisms include reversed phase, ion exchange, normal phase and mixed mode
  • 24 phases and counting
  • Cleaner extracts and reproducible results
TRUSTED_SUSTAINABILITY
Spécification
À utiliser avec (équipement) Vacuum Manifolds or Automated SPE Equipment
Granulométrie 20 μm
Adsorbant C18-E
Type Solid Phase Extraction Column