Learn More
R&D Systems™ Streptavidin-Fluorescein
Code produit 18401580
Modifier l'affichage
Click to view available options
Quantité:
30 μL
Conditionnement:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18401580 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18401580 Fournisseur R&D Systems™ Code fournisseur 48003014
  • ONLINE EXCLUSIVE PRICE
    152.70€
    135.45€ / 30µL
    Économisez 17.25€
    11% de réduction
 Article restreint
Expédition estimée: 04-12-2026
For use with apoptosis detection reagents and kits
TRUSTED_SUSTAINABILITY
Spécification
Quantité 30 μL
Contenu et stockage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Type de produit Streptavidin-Fluorescein
Gamme de produits R&D Systems