Learn More
Sulfaphenazole, MedChemExpress
Code produit 16408696
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantité:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16408696 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16408696 Fournisseur Medchem Express Code fournisseur HYB1218
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Sulfaphenazole is a specific inhibitor of CYP2C9 which blocks atherogenic and pro-inflammatory effects of linoleic acid (increase in oxidative stress and activation of AP-1) mediated by CYP2C9.
Sulfaphenazole is a specific inhibitor of CYP2C9 which blocks atherogenic and pro-inflammatory effects of linoleic acid (increase in oxidative stress and activation of AP-1) mediated by CYP2C9.
Spécification
Nom chimique ou matériau Sulfaphenazole
Forme physique Powder
CAS 526-08-9
Couleur Blanc
Quantité 100 mg
Notes de qualité de la pureté Research
Formule moléculaire C15H14N4O2S
À utiliser avec (application) COVID-19-immunoregulation
Informations sur la solubilité DMSO : ≥ 100 mg/mL (318.11 mM)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=CC=NN2C3=CC=CC=C3)=O
Poids moléculaire (g/mol) 314.36
Poids de la formule 314.36
Pourcentage de pureté 98.0%
Qualité Research