Learn More
Thermo Scientific™ SureTect™ Staphylococcus aureus PCR Assay
Code produit 17855397
Code produit 17855397 Fournisseur Thermo Scientific™ Code fournisseur A44255
Rapidly detect Staphylococcus aureus in food and environmental samples with the Thermo ScientificTM SureTectTM Staphylococcus aureus PCR Assay.
Rapidly detect Staphylococcus aureus in food and environmental samples with the Thermo Scientific™ SureTect™ Staphylococcus aureus PCR Assay.The assay incorporates an optimized liquid lysis step, providing a quick and simple method for preparing the sample for rapid detection on the Applied Biosystems™7500 Fast Food Safety Real-Time PCR Systems or Applied Biosystems™ QuantStudio™ 5 Food Safety Real Time PCR System.The assay enables users to reliably detect and differentiate Staphylococcus aureus from food, including infant formula, and environmental samples in less than 24 hours.
The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.
Simple
  • Easy-to-use with single enrichment step and pre-dispensed reagents.
  • Detect multiple targets in the same run with universal PCR protocols across the range.
Fast
  • Optimized workflow delivers on-screen results in as little as 24 hours
  • Maximum flexibility for operational efficiency with the ability to run all assays simultaneously and to test up to 96 samples in each run.
Smart
  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation.
  • Actionable data supports fast decisions in a complex product supply chain.
The Thermo Scientific™ SureTect™ Staphylococcus aureus PCR Assay includes:
  • SureTect Lysis Reagent 1 Tubes
  • SureTect Lysis Tube Caps
  • SureTect Proteinase K
  • SureTect Staphylococcus aureus PCR Tubes
  • SureTect PCR Caps
Enriched samples are pipetted into pre-filled lysis tubes, along with Proteinase K, before incubation, to lyse any bacterial cells present in the sample and release their DNA into the solution. The lysate is then loaded into the SureTect Staphylococcus aureus PCR tube to re-hydrate the pre-loaded PCR pellet which contains all the necessary components and reagents for PCR, including a probe, primers and DNA template for the internal amplification control (IAC). The PCR tubes are then sealed, loaded into Applied Biosystems™ QuantStudio™ 5 Food Safety Real Time PCR instrument with RapidFinder™ Analysis Software v1.1 or later or the Applied Biosystems 7500 Fast Food Safety 96-well Real-Time PCR instrument, and the run started using the RapidFinder™ Express Software v2.0 or later. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed or downloaded as required. The internal amplification control confirms correct assay conditions have occurred.
As with all test methods, the source, formulation and quality of enrichment medium can influence the results. Thermo Fisher Scientific has evaluated the SureTect PCR Assays workflows with industry leading Thermo Scientific™ Oxoid™ and Thermo Scientific™ Remel™ media formulations, as well as, ISO formulations where fit for purpose.
Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY
Spécification
Description SureTect Staphylococcus aureus PCR Assay
Méthode de détection Amorce-sonde
Forme Liquide
GC-Rich PCR Performance Élevé
Vitesse de réaction Fast
À utiliser avec (équipement) Applied Biosystems 7500 Fast Real-Time Food Safety PCR instrument and Applied BiosystemsTM QuantStudioTM 5 Food Safety Real Time PCR SystemApplied Biosystems SimpliAMPRapidFinder Express v2.0 RapidFinder Analysis Software v.1.1
Nbre de tests 96 tests / kit
Type de produit Staphylococcus aureus PCR Assay