Learn More
MedChemExpress TAK-448 acetate
Code produit 16491798
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16491798 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16491798 Fournisseur MedChemExpress Code fournisseur HYP0076A
MedChemExpress TAK-448 acetate (MVT-602 acetate) is a potent and full KISS1R agonist with an IC50 of 460 pM and an EC50 of 632 pM.
Spécification
Pureté 99.37%
CAS 1470374-22-1
Contenu et stockage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
À utiliser avec (application) Cancer-programmed cell death
Poids moléculaire 1285.41 g/mol
Type de produit Peptides
Quantité 5 mg
Conditions d’expédition Room temperature
Solubilité DMSO : ≥ 100 mg/mL (77.80 mM) ∣H2O : 50 mg/mL (38.90 mM∣Need ultrasonic)
Synonyme MVT-602 acetate