Learn More
Talmapimod, MedChemExpress
Artikelnummer. p-200100866
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantité
16443948 10 mg
16432926 25 mg
16426667 5 mg
16400307 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443948 Supplier Medchem Express Supplier No. HY10406
This item is not returnable. View return policy
MedChemExpress Talmapimod (SCIO-469) is an orally active, selective, and ATP-competitive p38α inhibitor with an IC50 of 9 nM. Talmapimod shows about 10-fold selectivity over p38β, and at least 2000-fold selectivity over a panel of 20 other kinases, including other MAPKs.
Talmapimod (SCIO-469) is an orally active, selective, and ATP-competitive p38α inhibitor with an IC50 of 9 nM. Talmapimod shows about 10-fold selectivity over p38β, and at least 2000-fold selectivity over a panel of 20 other kinases, including other MAPKs.
Specifications
Nom chimique ou matériau Talmapimod
CAS 309913-83-5
Couleur Rose
Notes de qualité de la pureté Research
Formule moléculaire C27H30ClFN4O3
Quantité 10 mg
Synonyme SCIO-469
Informations sur la solubilité DMSO : ≥ 100 mg/mL (194.93 mM)
SMILES O=C(N(C)C)C(C1=CN(C)C2=C1C=C(C(N3[C@H](C)CN(CC4=CC=C(F)C=C4)[C@@H](C)C3)=O)C(Cl)=C2)=O
Poids moléculaire (g/mol) 513.0
Poids de la formule 513.0
Pourcentage de pureté 98.04%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-Kinase/protease