Learn More
Applied Biosystems™ TaqMan™ Microbe or ABR Assay
Code produit 17275996 Tous les produits Applied Biosystems Produits
Modifier l'affichage
Click to view available options
Quantité:
360 μl
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
17275996 360 μl 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17275996 Fournisseur Applied Biosystems™ Code fournisseur A50133
 Article restreint
Expédition estimée: 18-02-2027
Les dosages Applied Biosystems TaqMan Microbe Detection et TaqMan Antibiotic Resistance sont utilisés pour la détection quantitative en temps réel par PCR des espèces microbiennes et des gènes de résistance aux antibiotiques.
Applied Biosystems TaqMan Microbe Detection and TaqMan Antibiotic Resistance assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5′ end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3′ end.
Features of TaqMan Microbe Detection and TaqMan Antibiotic Resistance assays include:
  • Easy to use—just add DNA and master mix and run the qPCR—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low-abundance microbial targets
  • Accurate—identify microbial targets with high accuracy of quantitation
  • Extensive content—over 600 predesigned microbe detection assays and more than 130 predesigned antibiotic resistance assays available
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for closely related microbial species and subtypes
  • Fast delivery—assays shipped in as little as 1–2 days (see below)
Approximate ship time
  • Inventoried: 1–2 days in North America*
  • Made to order: 4–6 days in North America
All of our predesigned TaqMan gene expression assays are covered by the TaqMan Assays qPCR Guarantee.**
*Our most popular assays are stocked in North America for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.
**Subject to terms and conditions.
TRUSTED_SUSTAINABILITY
Spécification
Quantité 360 μl