Learn More
MedChemExpress Taspoglutide
Code produit 16463986
Encompass_Preferred
Change view
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16463986 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16463986 Fournisseur MedChemExpress Code fournisseur HYP0165
MedChemExpress Taspoglutide is a long-acting glucagon-like peptide 1 (GLP-1) receptor agonist developed for treatment of type 2 diabetes, with an EC50 value of 0.06 nM.
Spécification
Pureté 96.85%
CAS 275371-94-3
Contenu et stockage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
À utiliser avec (application) Metabolism-sugar/lipid metabolism
Poids moléculaire 3339.71 g/mol
Type de produit Peptides
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité DMSO : ≥ 28 mg/mL (8.38 mM)
Synonyme ITM077∣ R1583∣ BIM51077