Learn More
Tazemetostat, MedChemExpress
Code produit 16468517
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
200 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16468517 5 mg
16442017 50 mg
16435868 10 mg
16426336 100 mg
16471106 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Code produit 16468517 Fournisseur Medchem Express Code fournisseur HY13803
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Tazemetostat (EPZ-6438) is a potent, selective and orally available EZH2 inhibitor.
Tazemetostat (EPZ-6438) inhibits the activity of human polycomb repressive complex 2 (PRC2)-containing wild-type EZH2 with a Ki value of 2.5 nM. Tazemetostat (EPZ-6438) inhibits EZH2 with IC50s of 11 and 16 nM in peptide assay and nucleosome assay, respectively. Tazemetostat (EPZ-6438) inhibits rat EZH2 with an IC50 of 4 nM. Tazemetostat (EPZ-6438) also inhibits EZH1 with an IC50 of 392 nM.
Spécification
Nom chimique ou matériau Tazemetostat
Forme physique Solid
CAS 1403254-99-8
Couleur Jaune clair
Quantité 5 mg
Notes de qualité de la pureté Research
Synonyme EPZ-6438 E-7438
Informations sur la solubilité DMSO : 50 mg/mL (87.30 mM; Need ultrasonic) ∣0.1 M HCL : 14.29 mg/mL (24.95 mM; ultrasonic and adjust pH to 5 with HCl)
SMILES O=C(C1=CC(C2=CC=C(CN3CCOCC3)C=C2)=CC(N(CC)C4CCOCC4)=C1C)NCC5=C(C)C=C(C)NC5=O
Poids moléculaire (g/mol) 572.74
Pourcentage de pureté 98.0%
Qualité Research