Learn More
R&D Systems™ Terbinafine hydrochloride
Description
Terbinafine hydrochloride is a selective TASK3 activator (pEC50 = 6.2). Exhibits >10-fold selectivity for TASK3 over TREK2, TRESK, THIK1 and TASK2. Also inhibits TWIK1 (pIC50 = 5.69). Also antifungal; inhibits fungal sterol biosynthesis by inhibition of squalene epoxidase.
Spécification
Spécification
| CAS | 78628-80-5 |
| Synonyme | N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride |
| Quantité | 250 mg |
| Gamme de produits | Tocris |
| Formule moléculaire | C21H25N.HCl |
| Poids de la formule | 327.89 |
| Pureté | 98% |