Learn More
Thermo Scientific™ Thermo Scientific™ Ramsey™ Oretronic IV Tramp Metal Detector
Code produit 18882445 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Description:
Oretronic IV Tramp Metal Detector, 4 Control
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Description
18882445 Oretronic IV Tramp Metal Detector, 4 Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 18882445 Fournisseur Thermo Scientific™ Code fournisseur ORETRONICIV4
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
Règlementaire : Cet article est soumis à éco-contribution.
Minimize lost production time with an economical and reliable means to protect expensive crushers, conveyors and other process equipment from damage by tramp metal.
This product has been discontinued – effective June 26, 2023. Aftermarket support including remote technical support and some field service will continue to be available as well as spare parts based on material availability.
Please check for more details on the discontinuation and contact our service team here.
Ramsey Oretronic Tramp Metal Detector Features:
  • Microprocessor-based control unit automates system set-up and calibration.
  • Operator interface provides easy-to-read indicators and simplified setup and system maintenance.
  • Variable frequency eliminates interference from other electrical equipment.
  • Quick Balance electronically balances the coil with the push of a key.
  • Impact-and moisture-resistant PVC coils are mounted on a fiberglass support structure.
  • Offset Swing Arm protects the coils and reduces breakage.
  • Simple installation – no belt cutting, special tools or instruments.
  • Single coil (optional): A single, under-the-belt coil assembly eliminates damage from high burden depths and interference from overhead metal objects.
  • Distance indication (optional): When incorporating a speed sensor, the tramp metal detector’s display shows the distance from the coil to the tramp metal in tenths of feet.
  • Serial communications (optional): Plug in RS-485 communications board.
Additional Features for the Oretronic IV 4A Control:
  • Improved sensitivity.
  • NEMA-4X enclosure.
  • External reset button.
  • Optional RS-485 communication card.
Additional Features for the Oretronic IV 6A Control
  • High sensitivity (note: can’t use superlatives)
  • Painted metal steel enclosure.
  • External reset button.
  • Optional RS-485 communication card.
  • Optional heater for low temperature applications.
Additional Features for the Oretronic III Tramp Metal Detector:
  • Serial communications (RS-485) available as standard to communicate with a remote PLC or remote front panel.
TRUSTED_SUSTAINABILITY
Specifications
Description Oretronic IV Tramp Metal Detector, 4 Control