Learn More
Thiostrepton, MedChemExpress
Code produit 16464517
Encompass_Preferred
Change view
Click to view available options
Quantité:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
16464517 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16464517 Supplier Medchem Express Supplier No. HYB0990
This item is not returnable. View return policy
MedChemExpress Thiostrepton is a thiazole antibiotic which selectively inhibits FOXM1. FOXM1 binds to YAP/TEAD complex. YAP/TEAD/FOXM1 complex binding at regulatory regions of genes governing cell cycle may impact cell proliferation.
Thiostrepton is a thiazole antibiotic which selectively inhibits FOXM1. FOXM1 binds to YAP/TEAD complex. YAP/TEAD/FOXM1 complex binding at regulatory regions of genes governing cell cycle may impact cell proliferation.
Specifications
Nom chimique ou matériau Thiostrepton
Forme physique Powder
CAS 1393-48-2
Couleur Blanc
Quantité 50 mg
Notes de qualité de la pureté Research
Formule moléculaire C72H85N19O18S5
À utiliser avec (application) COVID-19-immunoregulation
Informations sur la solubilité DMSO : 125 mg/mL (75.08 mM; Need ultrasonic)
SMILES O=C([C@@H](NC(C(NC([C@@H](NC(C(NC(C=C1)C2([H])O)[C@@H](C)CC)=O)C)=O)=C)=O)C)N[C@](C3=NC(C(NC(C(N/C(C4=N[C@]5([H])CS4)=C\C)=O)[C@H](O)C)=O)=CS3)(CCC(C6=NC(C(NC(C(NC(C(N)=O)=C)=O)=C)=O)=CS6)=N7)[C@@]7([H])C8=CSC([C@@]([C@H](OC(C9=CC([C@@H](O)C)=C1C2=N9)=O)C)([H])NC(C%10=CSC([C@](NC5=O)([H])[C@](C)(O)[C@H](O)C)=N%10)=O)=N8
Poids moléculaire (g/mol) 1664.89
Poids de la formule 1664.89
Pourcentage de pureté 95.0%
Qualité Research