Learn More
TPP™ Tissue Culture Flasks
Description
Surface activated, uniform flat growth surface with crystal clear transparency
- Surface activated, uniform flat growth surface
- Crystal clear transparency
- Geometry of neck reduces risk of media slop over
- Stable, slip-free stacking of multiple TPP flasks
- Highly visible bilateral marking area
- White and accurate volume graduations for optic control of the filling volume
Specifications
Specifications
| Description | Dimensions: 210 x 122 x 50 mm |
| Matériau | Polystyrene |
| Nbre par carton | 36 |
| Graduated | Oui |
| Forme du fond | Plat |
| Surface de culture | 150 cm² |
| Volume (métrique) de travail | 15 à 45 ml |