Learn More
Thermo Scientific Chemicals TCL grade 12 microns, gel de silice APS
Code produit 11315587
Change view
Click to view available options
Quantité:
50 g
250 g
1 kg
5 kg
Conditionnement:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11315587 250 g 250g
11395577 1 kg 1kg
11305587 50 g 50g
11385577 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 11315587 Fournisseur Thermo Scientific Chemicals Code fournisseur 042765.30
Demander un format en vrac ou personnalisé
TLC plate preparation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Spécification
Granulométrie 12 μm
Porosité 60 Å
Qualité TLC
Quantité 250 g
Aire de la surface (métrique) 500 à 600 m2/g
Type Gel de silice
Matériau Silice