Learn More
Tocris Bioscience™ Astressin
GREENER_CHOICE
Code produit 18025652
Modifier l'affichage
Click to view available options
Quantité:
500 μg
Conditionnement:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18025652 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18025652 Fournisseur Tocris Bioscience™ Code fournisseur 1606/500U
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
Potent CRF receptor antagonist
Astressin is a potent corticotropin-releasing factor (CRF) receptor antagonist (Ki values are 2, 1.5 and 1 nM at CRF1, CRF2α and CRF2β). Reduces ACTH secretion, blocks delayed gastric emptying and is neuroprotective in vivo.
TRUSTED_SUSTAINABILITY
Spécification
Quantité 500 μg