Learn More
Topiroxostat, MedChemExpress
Code produit 16456576
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16456576 100 mg
16446228 10 mg
16422357 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 16456576 Fournisseur Medchem Express Code fournisseur HY14874
MedChemExpress Topiroxostat (FYX-051) is a potent and orally active xanthine oxidoreductase (XOR) inhibitor with an IC50 value of 5.3 nM and a Ki value of 5.7 nM. Topiroxostat exhibits weak CYP3A4-inhibitory activity (18.6%). Topiroxostat has the potential for hyperuricemia treatment.
Topiroxostat (FYX-051) is a potent and orally active xanthine oxidoreductase (XOR) inhibitor with an IC50 value of 5.3 nM and a Ki value of 5.7 nM. Topiroxostat exhibits weak CYP3A4-inhibitory activity (18.6%). Topiroxostat has the potential for hyperuricemia treatment.
Spécification
Nom chimique ou matériau Topiroxostat
Forme physique Solid
CAS 577778-58-6
Couleur Blanc
Quantité 100 mg
Notes de qualité de la pureté Research
Formule moléculaire C13H8N6
À utiliser avec (application) Metabolism-sugar/lipid metabolism
Synonyme FYX-051
Informations sur la solubilité DMSO : 23.5 mg/mL (94.67 mM; Need ultrasonic and warming)
SMILES N#CC1=NC=CC(C2=NNC(C3=CC=NC=C3)=N2)=C1
Poids moléculaire (g/mol) 248.24
Poids de la formule 248.24
Pourcentage de pureté 99.63%
Qualité Research