Learn More
TP-10, MedChemExpress
Code produit 16496088
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16496088 10 mg
16482227 50 mg
16418697 5 mg
16416496 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16496088 Fournisseur Medchem Express Code fournisseur HY14550
MedChemExpress TP-10 is a selective PDE10A inhibitor with an IC50 value of 0.8 nM. TP-10 shows an antioxidant activity with IC50s of 31.72 and 16.04 μg/ml for DPPH and CUPRAC, respectively. TP-10 can be used for the research of neuropathy.
TP-10 is a selective PDE10A inhibitor with an IC50 value of 0.8 nM. TP-10 shows an antioxidant activity with IC50s of 31.72 and 16.04 μg/ml for DPPH and CUPRAC, respectively. TP-10 can be used for the research of neuropathy.
Spécification
Nom chimique ou matériau TP-10
CAS 898563-00-3
Couleur Orange
Notes de qualité de la pureté Research
Formule moléculaire C26H19F3N4O
Quantité 10 mg
Informations sur la solubilité DMSO : 100 mg/mL (217.18 mM; Need ultrasonic)
SMILES FC(F)(F)CN1C=C(C2=CC=NC=C2)C(C(C=C3)=CC=C3OCC4=NC5=CC=CC=C5C=C4)=N1
Poids moléculaire (g/mol) 460.45
Poids de la formule 460.45
Pourcentage de pureté 98.0%
Qualité Research
Forme physique Solid
À utiliser avec (application) COVID-19-immunoregulation