missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Benzyl-N-Boc-L-proline, 95%, Thermo Scientific™

Product Code. 15428657
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428657 250 mg 250mg
15418657 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428657 Supplier Thermo Scientific Alfa Aesar Supplier No. H52135.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153074-95-4
Molecular Formula C17H23NO4
Molecular Weight (g/mol) 305.374
MDL Number MFCD07784045
InChI Key JPNHKKRLLDYCIZ-KGLIPLIRSA-N
Synonym 2s,4r-4-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, boc-2s,4r-4-benzyl-pyrrolidine-2-carboxylic acid, boc-trans-4-benzyl-l-proline, boc-r-gamma-benzyl-l-proline, 2s,4r-4-benzyl-1-2-methylpropan-2-yl oxycarbonyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-benzyl pyrrolidine-2-carboxylic acid, 4beta-benzylpyrrolidine-1,2alpha-dicarboxylic acid 1-tert-butyl ester
PubChem CID 2761964
IUPAC Name (2S,4R)-4-benzyl-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)CC2=CC=CC=C2

Specifications

Melting Point 100°C to 102°C
Quantity 250 mg
Formula Weight 305.37
Percent Purity 95%
Chemical Name or Material trans-4-Benzyl-N-Boc-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.