Learn More
tri-2,4-xylyl Phosphate, TRC
Code produit 30009564
Change view
Click to view available options
Quantité:
100 mg
500 mg
1 g
Conditionnement:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30009564 500 mg 500mg
30005516 1 g 1g
18062910 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30009564 Fournisseur Toronto Research Chemicals Code fournisseur TRCT891900500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3862-12-2
Formule moléculaire C24H27O4P
Poids moléculaire (g/mol) 410.45
Synonyme Dimethyl-phenol Phosphate, Xylenol Phosphate, Xylyl Phosphate, Antiblaze 524, Antiblaze TXP, Coalite NTP, Durad 220X, Durad TXP, Fyrquel L, Phosflex 179, Phosphoric Acid Trixylyl Ester, TXP, Trixylenyl Phosphate, tris(2,4-dimethylphenyl) phosphate, Tri(2,4-xylenyl) phosphate
Nom IUPAC tris(2,4-dimethylphenyl) phosphate
SMILES O=P(OC1=CC=C(C=C1C)C)(OC2=CC=C(C=C2C)C)OC3=CC=C(C=C3C)C
Specifications
Nom chimique ou matériau Trixylyl Phosphate
Notes de qualité de la pureté HPLC
Quantité 500 mg
Formule InChI InChI=1S/C24H27O4P/c1-16-7-10-22(19(4)13-16)26-29(25,27-23-11-8-17(2)14-20(23)5)28-24-12-9-18(3)15-21(24)6/h7-15H,1-6H3
Poids de la formule 410.16
Pourcentage de pureté >95%