Learn More
Triptonide, MedChemExpress
Code produit 16428048
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16428048 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16428048 Fournisseur Medchem Express Code fournisseur HY32736
MedChemExpress Triptonide (NSC 165677) is a natural product identified in Tripterygium wilfordii Hook F.. Triptonide is a Wnt signaling inhibitor with an IC50 of appropriately 0.3 nM. Triptonide has immunosuppression, anti-inflammatory, anti-fertility, neuroprotective and anti-lymphoma effects.
Triptonide (NSC 165677) is a natural product identified in Tripterygium wilfordii Hook F.. Triptonide is a Wnt signaling inhibitor with an IC50 of appropriately 0.3 nM. Triptonide has immunosuppression, anti-inflammatory, anti-fertility, neuroprotective and anti-lymphoma effects.
Spécification
Nom chimique ou matériau Triptonide
CAS 38647-11-9
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C20H22O6
Quantité 10 mg
Synonyme NSC 165677 PG 492
Informations sur la solubilité DMSO : 100 mg/mL (279.03 mM; Need ultrasonic)
SMILES O=C([C@@]1([C@H]2O1)C(C)C)[C@]34O[C@H]3C[C@@]([H])(C5=C(CC6)C(OC5)=O)[C@@]6(C)[C@]47[C@H]2O7
Poids moléculaire (g/mol) 358.39
Poids de la formule 358.39
Pourcentage de pureté 98.63%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death